Hydrocodone
Synonym(s):
Hydrocodone, 4,5alpha-Epoxy-3-methoxy-17-methylmorphinan-6-one
Empirical Formula (Hill Notation):
C18H21NO3
CAS Number:
Molecular Weight:
299.36
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing