
Hydrobromic acid solution

Synonym(s):
Hydrogen bromide in acetic acid
Empirical Formula (Hill Notation):
HBr
Molecular Weight:
80.91
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing