Skip to Content
Merck
HEPES potassium salt ≥99.5% (titration)

HEPES potassium salt

HEPES potassium salt ≥99.5% (titration)
Synonym(s):
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid potassium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) potassium salt
Empirical Formula (Hill Notation):
C8H17KN2O4S
CAS Number:
Molecular Weight:
276.39

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing