Halcinonide
Synonym(s):
(11β, 16α)-21-Chloro-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregn-4-ene-3,20-dione
Empirical Formula (Hill Notation):
C24H32ClFO5
CAS Number:
Molecular Weight:
454.96
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing