
Glycocholic acid hydrate

Synonym(s):
3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid N-(carboxymethyl)amide, Cholylglycine, N-(3α,7α,12α-Trihydroxy-24-oxocholan-24-yl)-glycine
Empirical Formula (Hill Notation):
C26H43NO6 · xH2O
CAS Number:
Molecular Weight:
465.62 (anhydrous basis)
Products
Product Category
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Technique
Showing 1-1 of 1