Skip to Content
Merck
γ-Butyrolactone analytical standard

γ-Butyrolactone

γ-Butyrolactone analytical standard
Synonym(s):
γ-Hydroxybutyric acid lactone, 4-Hydroxybutyric acid lactone, GBL
Empirical Formula (Hill Notation):
C4H6O2
CAS Number:
Molecular Weight:
86.09
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing