Skip to Content
Merck
Gallium(III)-phthalocyanine chloride Dye content 97 %

Gallium(III)-phthalocyanine chloride

Gallium(III)-phthalocyanine chloride Dye content 97 %
Synonym(s):
Phthalocyanine gallium(III) monochloride salt
Empirical Formula (Hill Notation):
C32H16ClGaN8
CAS Number:
Molecular Weight:
617.70

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing