
Gadolinium(III) chloride solution

Linear Formula:
GdCl3
CAS Number:
Molecular Weight:
263.61
Products
Brand
Formula Weight
Format
Chemical Composition
Quality Segment
Reagent Type
Core
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing