Skip to Content
Merck
Fmoc-D-Trp-OH Novabiochem®

Fmoc-D-Trp-OH

Fmoc-D-Trp-OH Novabiochem®
Synonym(s):
Fmoc-D-Trp-OH
Empirical Formula (Hill Notation):
C26H22N2O4
CAS Number:
Molecular Weight:
426.46

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing