![Fluoxetine Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/183/732/fef0009b-c7cf-4a94-b991-6919a59a68d4/640/fef0009b-c7cf-4a94-b991-6919a59a68d4.png)
Fluoxetine Related Compound A
![Fluoxetine Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/structures/183/732/fef0009b-c7cf-4a94-b991-6919a59a68d4/640/fef0009b-c7cf-4a94-b991-6919a59a68d4.png)
Synonym(s):
(±)-N-Methyl-(γ-[3-(trifluoromethyl)phenoxy]benzenepropanamine hydrochloride
Empirical Formula (Hill Notation):
C17H18F3NO · HCl
CAS Number:
Molecular Weight:
345.79
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2