Fluorescein
Synonym(s):
Acid Yellow 73
Empirical Formula (Hill Notation):
C20H12O5
CAS Number:
Molecular Weight:
332.31
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Special Grade
Technique
Showing 1-3 of 3