Flumazenil Related Compound C
Synonym(s):
N,N-Dimethylformamide diethyl acetal, 1,1-Diethoxy-N,N-dimethylmethylamine, 1,1-Diethoxytrimethylamine
Linear Formula:
(CH3)2NCH(OC2H5)2
CAS Number:
Molecular Weight:
147.22
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2