![Fluconazole Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/297/792/ce88fe84-78c0-4a6e-a260-f19924e66e06/640/ce88fe84-78c0-4a6e-a260-f19924e66e06.jpg)
Fluconazole Related Compound C
![Fluconazole Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/297/792/ce88fe84-78c0-4a6e-a260-f19924e66e06/640/ce88fe84-78c0-4a6e-a260-f19924e66e06.jpg)
Synonym(s):
1,1′-(1,3-Phenylene)bis-1H-1,2,4-triazole
Empirical Formula (Hill Notation):
C10H8N6
CAS Number:
Molecular Weight:
212.21
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2