Fenofibrate impurity B
Synonym(s):
2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid, Fenofibric acid
Empirical Formula (Hill Notation):
C17H15ClO4
CAS Number:
Molecular Weight:
318.75
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing