Ethyl stearate
Synonym(s):
Ethyl octadecanoate, Stearic acid ethyl ester
Linear Formula:
CH3(CH2)16COOC2H5
CAS Number:
Molecular Weight:
312.53
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Functional Group
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing