Ethylenedinitrilotetraacetic acid dipotassium salt dihydrate
Synonym(s):
Ethylenedinitrilotetraacetic acid dipotassium salt dihydrate
Empirical Formula (Hill Notation):
C10H14K2N2O8 · 2H2O
CAS Number:
Molecular Weight:
404.45
Products
Brand
Formula Weight
Physical Form
Quality Segment
Reaction Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing