Ethylene-d4 oxide
Synonym(s):
Deuterated ethylene oxide, Oxirane-D4
Empirical Formula (Hill Notation):
C2D4O
CAS Number:
Molecular Weight:
48.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Isotope
Application
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing