Skip to Content
Merck
Ethyl 2,2-diphenylacetate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Ethyl 2,2-diphenylacetate

Ethyl 2,2-diphenylacetate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Diphenylacetic acid ethyl ester, Ethyl diphenylacetate
Empirical Formula (Hill Notation):
C16H16O2
CAS Number:
Molecular Weight:
240.30

Products

Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Application
Special Grade