Ethyl 1,3-dithiane-2-carboxylate
Synonym(s):
Glyoxylic acid ethyl ester trimethylenemercaptal
Empirical Formula (Hill Notation):
C7H12O2S2
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing