![EPPS BioXtra, ≥99.5% (titration)](/deepweb/assets/sigmaaldrich/product/structures/189/850/ecf8ccc6-e98f-4e6a-8639-e12ea6bf6f74/640/ecf8ccc6-e98f-4e6a-8639-e12ea6bf6f74.png)
EPPS
![EPPS BioXtra, ≥99.5% (titration)](/deepweb/assets/sigmaaldrich/product/structures/189/850/ecf8ccc6-e98f-4e6a-8639-e12ea6bf6f74/640/ecf8ccc6-e98f-4e6a-8639-e12ea6bf6f74.png)
Synonym(s):
4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid, 4-(2-Hydroxyethyl)piperazine-1-propanesulfonic acid, N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulfonic acid), HEPPS
Empirical Formula (Hill Notation):
C9H20N2O4S
Products
Product Category
Brand
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing