![Edetate disodium United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/177/279/de9a18cd-88da-498c-ad1f-ca6a67d22fab/640/de9a18cd-88da-498c-ad1f-ca6a67d22fab.png)
Edetate disodium
![Edetate disodium United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/177/279/de9a18cd-88da-498c-ad1f-ca6a67d22fab/640/de9a18cd-88da-498c-ad1f-ca6a67d22fab.png)
Synonym(s):
Ethylenediaminetetraacetic acid disodium salt dihydrate, Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2
Empirical Formula (Hill Notation):
C10H14N2Na2O8 · 2H2O
CAS Number:
Molecular Weight:
372.24
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing