Skip to Content
Merck
ECC ChromoSelect Selective Agar suitable for microbiology, NutriSelect® Plus

ECC ChromoSelect Selective Agar

ECC ChromoSelect Selective Agar suitable for microbiology, NutriSelect® Plus
Synonym(s):
E. coli-Coliform Selective Agar ChromoSelect, ECC Selective Agar ChromoSelect

Products

Product Category
Brand
pH Value
Physical Form
Chemical Composition
Quality Segment
Application
Microorganism Suitability
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing