Skip to Content
Merck
(+)-O,O′-Di-p-toluoyl-D-tartaric acid made from synthetic tartaric acid, 97%

(+)-O,O′-Di-p-toluoyl-D-tartaric acid

(+)-O,O′-Di-p-toluoyl-D-tartaric acid made from synthetic tartaric acid, 97%
Synonym(s):
Di-p-toluoyl-D-tartaric acid
Linear Formula:
[CH3C6H4CO2CH(CO2H)-]2
CAS Number:
Molecular Weight:
386.35
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing