
DMT-Biotin-C6 Phosphoramidite

Synonym(s):
5′ Biotin Amidite, 5′-Biotin Phosphoramidite, 6-[(2-Cyanoethoxy)diisopropylaminophosphanoxy]-[1-N-(4,4′-dimethoxytrityl)biotinyl]aminohexane, DMT-Biotin-C6 Amidite, N-DMT-Biotinyl-2-Aminoethoxyethanol
Empirical Formula (Hill Notation):
C46H64N5O6PS
CAS Number:
Molecular Weight:
846.07
Products
Product Category
Brand
Formula Weight
Product Line
Purity
Quality Segment
Showing 1-3 of 3