Di-tert-butyl acetylenedicarboxylate
Synonym(s):
2-Butynedioic acid di-tert-butyl ester, Di-tert-butyl 2-butynedioate
Linear Formula:
(CH3)3COCOC≡CCOOC(CH3)3
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing