Skip to Content
Merck
Diquafosol tetrasodium >95% (HPLC)

Diquafosol tetrasodium

Diquafosol tetrasodium >95% (HPLC)
Synonym(s):
Diuridine 5′-tetraphosphate, Up4U tetrasodium
Empirical Formula (Hill Notation):
C18H22N4Na4O23P4
CAS Number:
Molecular Weight:
878.23

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing