Diethylstilbestrol
Synonym(s):
(E)-3,4-Bis(4-hydroxyphenyl)-3-hexene, DES, Stilbestrol
Empirical Formula (Hill Notation):
C18H20O2
CAS Number:
Molecular Weight:
268.35
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing