Skip to Content
Merck
Dichloroprop-P PESTANAL®, analytical standard

Dichloroprop-P

Dichloroprop-P PESTANAL®, analytical standard
Synonym(s):
(R)-(+)-2-(2,4-Dichlorophenoxy)propionic acid
Empirical Formula (Hill Notation):
C9H8Cl2O3
CAS Number:
Molecular Weight:
235.06
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing