![(DHQD)2PHAL ≥95%](/deepweb/assets/sigmaaldrich/product/structures/152/702/7d9bb6ef-ee7e-4194-a3b8-817f1f34b015/640/7d9bb6ef-ee7e-4194-a3b8-817f1f34b015.png)
(DHQD)2PHAL
![(DHQD)2PHAL ≥95%](/deepweb/assets/sigmaaldrich/product/structures/152/702/7d9bb6ef-ee7e-4194-a3b8-817f1f34b015/640/7d9bb6ef-ee7e-4194-a3b8-817f1f34b015.png)
Synonym(s):
Hydroquinidine 1,4-phthalazinediyl diether
Empirical Formula (Hill Notation):
C48H54N6O4
CAS Number:
Molecular Weight:
778.98
Products
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing