Deuterium chloride solution
Synonym(s):
Hydrochloric acid-d solution
Linear Formula:
DCl
CAS Number:
Molecular Weight:
37.47
Products
Product Category
Brand
Formula Weight
Isotope
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing