![Delphinidin 3-glucoside chloride phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/208/527/e318371c-0ab7-486a-add6-1d817f52cdb9/640/e318371c-0ab7-486a-add6-1d817f52cdb9.png)
Delphinidin 3-glucoside chloride
![Delphinidin 3-glucoside chloride phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/208/527/e318371c-0ab7-486a-add6-1d817f52cdb9/640/e318371c-0ab7-486a-add6-1d817f52cdb9.png)
Synonym(s):
Delphinidin 3-O-β-D-glucoside chloride, Delphinidin 3-O-β-D-glucopyranoside chloride, Delphinidin 3-monoglucoside, Delphinin chloride, Myrtillin chloride
Empirical Formula (Hill Notation):
C21H21ClO12
CAS Number:
Molecular Weight:
500.84
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing