Skip to Content
Merck
δ-BHC certified reference material, 1000 μg/mL in methanol

δ-BHC

δ-BHC certified reference material, 1000 μg/mL in methanol
Synonym(s):
δ-HCH solution, δ-1,2,3,4,5,6-Hexachlorocyclohexane solution
Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing