![Dalfampridine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/318/333/7bac32ac-15d3-46b3-af6d-cf53c2e95bf3/640/7bac32ac-15d3-46b3-af6d-cf53c2e95bf3.png)
Dalfampridine
![Dalfampridine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/318/333/7bac32ac-15d3-46b3-af6d-cf53c2e95bf3/640/7bac32ac-15d3-46b3-af6d-cf53c2e95bf3.png)
Synonym(s):
4-Aminopyridine, 4-Pyridinamine, 4-Pyridylamine, 4AP, Fampridine
Empirical Formula (Hill Notation):
C5H6N2
CAS Number:
Molecular Weight:
94.11
Products
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing