![[D-Ala2]-Deltorphin II ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/289/271/db4c3fc4-8ba9-46ef-b214-158c9758871f/640/db4c3fc4-8ba9-46ef-b214-158c9758871f.png)
[D-Ala2]-Deltorphin II
![[D-Ala2]-Deltorphin II ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/289/271/db4c3fc4-8ba9-46ef-b214-158c9758871f/640/db4c3fc4-8ba9-46ef-b214-158c9758871f.png)
Synonym(s):
Tyr-D-Ala-Phe-Glu-Val-Val-Gly amide
Empirical Formula (Hill Notation):
C38H54N8O10
CAS Number:
Molecular Weight:
782.88
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1