(+)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene
Synonym(s):
(2S,2′S,5S,5′S)-2,2′,5,5′-Tetraethyl-1,1′-(o-phenylene)diphospholane, (S,S)-Ethyl-DUPHOS
Empirical Formula (Hill Notation):
C22H36P2
CAS Number:
Molecular Weight:
362.47
Products
Brand
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing