Skip to Content
Merck
Cytochalasin D, Zygosporium mansonii InSolution, ≥98%, 10 mM in DMSO, A cell-permeable fungal toxin

Cytochalasin D

Cytochalasin D, Zygosporium mansonii InSolution, ≥98%, 10 mM in DMSO, A cell-permeable fungal toxin
Synonym(s):
Zygosporin A, InSolution Cytochalasin D, Zygosporium mansonii
Empirical Formula (Hill Notation):
C30H37NO6
CAS Number:
Molecular Weight:
507.62

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action