Skip to Content
Merck
Cyclooctatetraene 98%

Cyclooctatetraene

Cyclooctatetraene 98%
Empirical Formula (Hill Notation):
C8H8
CAS Number:
Molecular Weight:
104.15

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing