
Cyanidin 3-rutinoside chloride

Synonym(s):
Keracyanin chloride, Cyanidin-3-O-rutinoside chloride
Empirical Formula (Hill Notation):
C27H31ClO15
CAS Number:
Molecular Weight:
630.98
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing