Skip to Content
Merck
Corticotropin Releasing Factor human, rat ≥95% (HPLC), powder

Corticotropin Releasing Factor human, rat

Corticotropin Releasing Factor human, rat ≥95% (HPLC), powder
Synonym(s):
CRF, CRH
Empirical Formula (Hill Notation):
C208H344N60O63S2
CAS Number:
Molecular Weight:
4757.45

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing