Coniferyl alcohol
Synonym(s):
3-(4-Hydroxy-3-methoxyphenyl)-2-propen-1-ol, 4-Hydroxy-3-methoxycinnamyl alcohol
Linear Formula:
HOC6H3(OCH3)CH=CHCH2OH
CAS Number:
Molecular Weight:
180.20
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reference Material Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing