Skip to Content
Merck
Codeine European Pharmacopoeia (EP) Reference Standard

Codeine

Codeine European Pharmacopoeia (EP) Reference Standard
Empirical Formula (Hill Notation):
C18H21NO3
CAS Number:
Molecular Weight:
299.36

Products

Product Category
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing