Skip to Content
Merck
Cobalt(II) 2,3-naphthalocyanine Dye content 85 %

Cobalt(II) 2,3-naphthalocyanine

Cobalt(II) 2,3-naphthalocyanine Dye content 85 %
Synonym(s):
2,3-Naphthalocyanine cobalt(II) salt
Empirical Formula (Hill Notation):
C48H24CoN8
CAS Number:
Molecular Weight:
771.69

Products

Brand
Formula Weight
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing