Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cobalt/Boron/Silicon/Iron/Molybdenum/Nickel
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF41999811
Cobalt/Boron/Silicon/Iron/Molybdenum/Nickel,
foil, Co 69%/B 12%/Si 12%/Fe 4%/Mo 2%/Ni 1%, thickness 0.025 mm, length 2 m, coil width 50 mm
Expand
View Pricing
GF49282689
Cobalt/Boron/Silicon/Iron/Molybdenum/Nickel,
foil, Co 69%/B 12%/Si 12%/Fe 4%/Mo 2%/Ni 1%, thickness 0.025 mm, length 0.5 m, coil width 50 mm
Expand
View Pricing
GF82593287
Cobalt/Boron/Silicon/Iron/Molybdenum/Nickel,
foil, thickness 0.025 mm, Co69%/B 12%/Si12%/Fe 4%/Mo 2%/Ni 1%, length 20 m, coil width 50 mm
Expand
View Pricing
GF60335995
Cobalt/Boron/Silicon/Iron/Molybdenum/Nickel,
foil, thickness 0.025 mm, Co69%/B 12%/Si12%/Fe 4%/Mo 2%/Ni 1%, coil width 50 mm, length 1 m
Expand
View Pricing
GF47060748
Cobalt/Boron/Silicon/Iron/Molybdenum/Nickel,
foil, Co 69%/B 12%/Si 12%/Fe 4%/Mo 2%/Ni 1%, thickness 0.025 mm, length 10 m, coil width 50 mm
Expand
View Pricing
GF49976487
Cobalt/Boron/Silicon/Iron/Molybdenum/Nickel,
foil, Co 69%/B 12%/Si 12%/Fe 4%/Mo 2%/Ni 1%, thickness 0.025 mm, length 5 m, coil width 50 mm
Expand
View Pricing