Skip to Content
Merck
Clorazepate dipotassium United States Pharmacopeia (USP) Reference Standard

Clorazepate dipotassium

Clorazepate dipotassium United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Clorazepate dipotassium salt, 7-Chloro-2,3-dihydro-2,2-dihydroxy-5-phenyl-1H-1,4-benzodiazepine-3-carboxylic acid
Linear Formula:
C16H11ClN2O4K2
CAS Number:
Molecular Weight:
408.92

Products

Brand
Feature
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing