Skip to Content
Merck
Clemizole hydrochloride ≥98% (HPLC)

Clemizole hydrochloride

Clemizole hydrochloride ≥98% (HPLC)
Synonym(s):
1-(p-Chlorobenzyl)-2-(1-pyrrolidinylmethyl)benzimidazole hydrochloride, 1-(p-Chlorobenzyl)-2-pyrrolidylmethylenebenzimidazole hydrochloride, 1-[(4-Chlorophenyl)methyl]-2-(1-pyrrolidinylmethyl)-1H-benzimidazole hydrochloride, CID 2782 hydrochloride, NSC 46261 hydrochloride
Empirical Formula (Hill Notation):
C19H20ClN3 · HCl
CAS Number:
Molecular Weight:
362.30
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing