![Clarithromycin Related Compound A United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/293/247/4d90cd96-9bbd-40c3-bd75-0bde628885b1/640/4d90cd96-9bbd-40c3-bd75-0bde628885b1.png)
Clarithromycin Related Compound A
![Clarithromycin Related Compound A United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/293/247/4d90cd96-9bbd-40c3-bd75-0bde628885b1/640/4d90cd96-9bbd-40c3-bd75-0bde628885b1.png)
Synonym(s):
6,11-Di-O-methylerythromycin A
Empirical Formula (Hill Notation):
C39H71NO13
CAS Number:
Molecular Weight:
761.98
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing