Skip to Content
Merck
CL 316,243 hydrate ≥98% (HPLC), powder

CL 316,243 hydrate

CL 316,243 hydrate ≥98% (HPLC), powder
Synonym(s):
Disodium 5-[(2R)-2-[[(2R)-2-(3-Chlorophenyl)-2-hydroxyethyl]amino]propyl]-1,3-benzodioxole-2,2-dicarboxylate hydrate
Empirical Formula (Hill Notation):
C20H18ClNNa2O7 · xH2O
CAS Number:
Molecular Weight:
465.79 (anhydrous basis)

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing