cis-Vaccenic acid
Synonym(s):
cis-11-Octadecenoic acid
Linear Formula:
CH3(CH2)5CH=CH(CH2)9CO2H
CAS Number:
Molecular Weight:
282.46
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Tissue
Functional Group
Application
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing