cis-Dichlorobis(triphenylphosphine)platinum(II)
Synonym(s):
cis-Bis(triphenylphosphine)platinum(II) dichloride
Linear Formula:
[(C6H5)3P]2PtCl2
CAS Number:
Molecular Weight:
790.56
Products
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing