
cis-4-Amino-cyclohexanol hydrochloride

Synonym(s):
cis-4-Hydroxycyclohexylamine hydrochloride
Empirical Formula (Hill Notation):
C6H14ClNO
Molecular Weight:
151.63
Products
Product Category
Brand
Formula Weight
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing